Learn More
PAK1 (phospho T212), Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic phosphopeptide of PAK1.
Supplier: Abnova Corporation PAB0569
Description
PAK proteins are critical effectors that link RhoGTPases to cytoskeleton reorganization and nuclear signaling. PAK proteins, a family of serine/threonine p21-activating kinases, include PAK1, PAK2, PAK3 and PAK4. These proteins serve as targets for the small GTP binding proteins Cdc42 and Rac and have been implicated in a wide range of biological activities. PAK1 regulates cell motility and morphology. Alternativelt spliced transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq
Specifications
PAK1 | |
Polyclonal | |
Rabbit polyclonal antibody raised against synthetic phosphopeptide of PAK1. | |
In PBS (0.09% sodium azide) | |
MGC130000/MGC130001/PAKalpha | |
Rabbit | |
Protein A purification | |
RUO | |
5058 | |
Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Dot Blot, ELISA | |
Unconjugated | |
Dot Blot (1:500) The optimal working dilution should be determined by the end user. | |
PAK1 | |
PAK1 | |
Synthetic phosphopeptide (conjugated with KLH) corresponding to residues surrounding T212 of human PAK1. | |
400 μL | |
Primary | |
Human | |
Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only