Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PAK2 Rabbit anti-Human, Mouse, Rabbit, Rat, Polyclonal Antibody, Abnova™

Catalog No. 89134004 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-134-004 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-134-004 Supplier Abnova Corporation Supplier No. PAB8890

Rabbit polyclonal antibody raised against synthetic peptide of PAK2.

The p21 activated kinases (PAK) are critical effectors that link Rho GTPases to cytoskeleton reorganization and nuclear signaling. The PAK proteins are a family of serine/threonine kinases that serve as targets for the small GTP binding proteins, CDC42 and RAC1, and have been implicated in a wide range of biological activities. The protein encoded by this gene is activated by proteolytic cleavage during caspase-mediated apoptosis, and may play a role in regulating the apoptotic events in the dying cell. [provided by RefSeq

Specifications

Antigen PAK2
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description p21 protein (Cdc42/Rac)-activated kinase 2
Formulation In PBS (30% glycerol, 0.5% BSA, 0.01% thimerosal)
Gene PAK2
Gene Alias PAK65, PAKgamma
Gene Symbols PAK2
Host Species Rabbit
Immunogen A synthetic peptide corresponding to residues surrounding amino acids 5 of human PAK2.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5062
Test Specificity This antibody recognizes 60kDa Pak2. The antibosy does not cross-react with Pak1, Pak3, and Pak4-6.
Target Species Human, Mouse, Rabbit, Rat
Content And Storage Store at -20°C. For long term storage store at -80°C. Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.