Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pak6, Rabbit, Polyclonal Antibody, Abnova™

Catalog No. 89119577 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-119-577 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-119-577 Supplier Abnova Corporation Supplier No. PAB17957

Rabbit polyclonal antibody raised against synthetic peptide of Pak6.

Specifications

Antigen Pak6
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description p21 (CDKN1A)-activated kinase 6
Formulation In PBS pH 7.2 (30% glycerol, 0.5% BSA, 0.01% thimerosal)
Gene Pak6
Gene Alias 4732456M09
Gene Symbols Pak6
Host Species Rabbit
Immunogen A synthetic peptide corresponding to the residues surrounding amino acid 552 of mouse Pak6.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 214230
Test Specificity This antibody recognizes ∽75kDa of Pak6 in Western Blot.
Target Species Equine, Human, Mouse, Pig, Rat
Content And Storage Store at -20°C. For long term storage store at -80°C. Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.