Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Rabbit anti-Pancreatic Lipase/PNLIP, Polyclonal,

Catalog No. ABS547MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ABS547MI 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ABS547MI Supplier MilliporeSigma™ Supplier No. ABS547
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Specifically detects Pancreatic Lipase/PNLIP in Human, Mouse, Rat samples, and it is validated for Western Blotting

Pancreatic triacylglycerol lipase (EC 3.1.1.3; UniProt P35638; also known as Pancreatic lipase, PL, PTL, Triacylglycerol acylhydrolase) is encoded by the PNLIP (Also known as PNLIPD) gene in human (Gene ID 5406). PTL is a pancreas synthesized carboxyl esterase that hydrolyzes insoluble, emulsified triglycerides (TAGs). The dietary TAGs are partially digested by the gastric lipase in the stomach. The emulsion particles then mix with digestive enzymes from the pancreas in the duodenum where digestion proceeds. The pancreas secretes multiple lipases, including pancreatic triglyceride lipase (PTL), carboxyl ester lipase (CEL), and pancreatic lipase-related protein-1 and -2 (PLRP1 & PLRP2). Unlike PLRP2, PLRP1 exhibits little activity against triglycerides and no measurable activity against phospholipids, galactolipids or cholesterol esters. Experimental evidences suggest that PLRP1 plays a negative regulatory role against PLT–colipase-mediated dietary triglyceride digestion. Pancreas-derived lipases are essential for the efficient digestion of dietary TAG. Pancreatic insufficiency or exocrine pancreas failure results in steatorrhea or steatorrhoea, where increased fat excretion is seen in fecal samples.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Pancreatic Lipase/PNLIP
Applications Western Blot
Classification Polyclonal
Formulation Rabbit polyclonal serum with 0.05% sodium azide.
Gene Accession No. P16233
Gene Symbols PNLIP
Host Species Rabbit
Immunogen Recombinant protein corresponding to human Pancreatic Lipase/PNLIP.
Purification Method Unpurified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Signaling
Primary or Secondary Primary
Gene ID (Entrez) NP_000927
Target Species Human, Mouse, Rat
Content And Storage Stable for one year at -20°C from date of receipt. Handling Recommendations: Upon receipt and prior to removing the cap centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles which may damage IgG and affect product performance.
Form Serum
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.