Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Rabbit anti-PAR-4, Polyclonal,

Catalog No. ABC502MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ABC502MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ABC502MI Supplier MilliporeSigma™ Supplier No. ABC502
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Specifically detects PAR-4 in Human samples, and it is validated for Western Blotting

PAR-4 is a pro-apoptotic protein capable of selectively inducing apoptosis in cancer cells and sensitizing the cells to diverse apoptotic stimuli causing tumor regression. PAR-4 also seems to be a transcriptional repressor and is involved in regulating APP expression and the cleavage activity of BACE1. PAR-4 interacts with several proteins including WT1, but also various kinases, and structural proteins like Actin, BACE1, SPSB1, SPSB2, and SPSB4. PAR4 is localized in the cytoplasm in non-apoptotic cells but is nuclear localized in cancer and apoptotic cells. PAR-4 is widely expressed and elevated in neurodegenerative diseases such as ALS, Alzheimer, Parkinson and Huntington diseases as well as stroke. PAR-4 is down regulated in cancers.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PAR-4
Applications Western Blot
Classification Polyclonal
Formulation Purified rabbit polyclonal in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide.
Gene Accession No. Q96IZ0
Gene Symbols PAWR; PAR4
Host Species Rabbit
Immunogen GST-tagged recombinant protein corresponding to the C-terminus of human PAR-4.
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Apoptosis, Cancer
Primary or Secondary Primary
Gene ID (Entrez) NP_002574
Target Species Human
Content And Storage Stable for one year at 2°C to 8°C from date of receipt.
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.