Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Rabbit anti-PAS Kinase, Polyclonal,

Catalog No. ABS1016MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ABS1016MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ABS1016MI Supplier MilliporeSigma™ Supplier No. ABS1016
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Specifically detects PAS Kinase in Human, Rat samples, and it is validated for Immunohistochemistry, Western Blotting

PAS domain-containing serine/threonine-protein kinase (EC 2.7.11.1; UniProt Q96RG2; also known as hPASK, PASKIN, PAS-kinase, Per-arnt-sim domain kinase) is encoded by the PASK (Also known as KIAA0135, STK37) gene in human (Gene ID 23178). PAS-kinase is a serine/threonine-protein kinase involved in energy homeostasis. Hepatic PASK expression is shown to be induced upon feeding and insulin stimulation, and PASK is reported to be essential for the proteolytic maturation of the transcription factor SREBP-1c in cultured cells and in the mouse and rat liver. SREBP-1c hyperactivation leads to excess hepatic fatty acids and triglycerides synthesis, which has been implicated as a causal feature of metabolic syndrome. PASK inhibition improves lipid and glucose metabolism in dietary animal models of obesity and dyslipidemia. PASK is also involved in protein translation regulation and is shown to phosphorylate eukaryotic translation elongation factor eEF1A1.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PAS Kinase
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Formulation Purified rabbit polyclonal antibody in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide.
Gene Accession No. Q96RG2
Gene Symbols PASK; KIAA0135
Host Species Rabbit
Immunogen KLH-conjugated linear peptide corresponding to human PAS Kinase near the C-terminus.
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Signaling
Primary or Secondary Primary
Gene ID (Entrez) NP_055963
Target Species Human, Rat
Content And Storage Stable for one year at 2°C to 8°C from date of receipt.
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.