Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PAX2 Rabbit anti-Mouse, Rat, Polyclonal Antibody, Abnova™

Catalog No. 89126375 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-126-375 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-126-375 Supplier Abnova Corporation Supplier No. PAB24709
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against synthetic peptide of PAX2.

PAX2 encodes paired box gene 2, one of many human homologues of the Drosophila melanogaster gene prd. The central feature of this transcription factor gene family is the conserved DNA-binding paired box domain. PAX2 is believed to be a target of transcriptional supression by the tumor supressor gene WT1. Mutations within PAX2 have been shown to result in optic nerve colobomas and renal hypoplasia. Alternative splicing of this gene results in multiple transcript variants. [provided by RefSeq

Specifications

Antigen PAX2
Applications Immunofluorescence, Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration 0.75-0.95 mg/mL
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against synthetic peptide of PAX2.
Dilution Western Blot (1:500) The optimal working dilution should be determined by the end user.
Formulation In Tris-glycine buffer, pH 7.4 (stabilizing proteins, cryogenic stabilizers, glycerol, 0.02% sodium azide)
Gene PAX2
Gene Symbols PAX2
Host Species Rabbit
Immunogen A synthetic peptide corresponding to amino acids 200-250 of human PAX2.
Purification Method Affinity Purification
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5076
Target Species Mouse, Rat
Content And Storage Store at -20°C
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.