Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PCSK9, Rabbit, Polyclonal Antibody, Abnova™

Catalog No. 89118834 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-118-834 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-118-834 Supplier Abnova Corporation Supplier No. PAB17045
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against full-length recombinant PCSK9.

This gene encodes a proprotein convertase belonging to the proteinase K subfamily of the secretory subtilase family. The encoded protein is synthesized as a soluble zymogen that undergoes autocatalytic intramolecular processing in the endoplasmic reticulum. The protein may function as a proprotein convertase. This protein plays a role in cholesterol homeostasis and may have a role in the differentiation of cortical neurons. Mutations in this gene have been associated with a third form of autosomal dominant familial hypercholesterolemia (HCHOLA3). [provided by RefSeq

Specifications

Antigen PCSK9
Applications Immunoprecipitation, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against full length recombinant PCSK9.
Dilution Western Blot (0.5-1 ug/mL) Immunoprecipitation (1-2 ug/sample) The optimal working dilution should be determined by the end user.
Formulation In 10mM phosphate buffer, 150mM NaCl, pH 7.5 (50% glycerol)
Gene PCSK9
Gene Alias FH3/HCHOLA3/LDLCQ1/NARC-1/NARC1
Gene Symbols PCSK9
Host Species Rabbit
Immunogen Recombinant His fusion protein corresponding to full length human PCSK9.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 255738
Target Species Human
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.