Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PDE11A Rabbit anti-Bovine, Chicken, Hamster, Horse, Human, Monkey, Mouse, Pig, Rabbit, Rat, Zebra fish, Polyclonal Antibody, Abnova™

Catalog No. 89118315 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-118-315 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-118-315 Supplier Abnova Corporation Supplier No. PAB16443
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against synthetic peptide of PDE11A.

The 3',5'-cyclic nucleotides cAMP and cGMP function as second messengers in a wide variety of signal transduction pathways. 3',5'-cyclic nucleotide phosphodiesterases (PDEs) catalyze the hydrolysis of cAMP and cGMP to the corresponding 5'-monophosphates and provide a mechanism to downregulate cAMP and cGMP signaling. This gene encodes a member of the PDE protein superfamily. Mutations in this gene are a cause of Cushing disease and adrenocortical hyperplasia. Multiple transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq]

Specifications

Antigen PDE11A
Applications Immunohistochemistry (PFA fixed)
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against synthetic peptide of PDE11A.
Dilution The optimal working dilution should be determined by the end user.
Formulation In PBS (0.09% sodium azide)
Gene PDE11A
Gene Alias FLJ23693/MGC133355/MGC133356/PPNAD2
Gene Symbols PDE11A
Host Species Rabbit
Immunogen A synthetic peptide (conjugated with KLH) corresponding to human PDE11A.
Purification Method Immunoaffinity purification
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 50940
Test Specificity Internal domain of human.
Target Species Bovine, Chicken, Equine, Hamster, Human, Monkey, Mouse, Pig, Rabbit, Rat, Zebrafish
Content And Storage Store at 4°C. For long term storage store at -80°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.