Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pde3b, Rabbit, Polyclonal Antibody, Abnova™

Catalog No. 89126382 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-126-382 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-126-382 Supplier Abnova Corporation Supplier No. PAB24717
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against synthetic peptide of Pde3b.

Specifications

Antigen Pde3b
Applications Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 0.65 mg/mL
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against synthetic peptide of Pde3b.
Dilution WB (1:500) Immunostain (1:200) The optimal working dilution should be determined by the end user.
Formulation In Tris-glycine buffer, pH 7.4 (stabilizing proteins, cryogenic stabilizers, glycerol, 0.02% sodium azide)
Gene Pde3b
Gene Symbols Pde3b
Host Species Rabbit
Immunogen A synthetic peptide corresponding to rat Pde3b.
Purification Method Affinity Purification
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 29516
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.