Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pde4dip, Rabbit, Polyclonal Antibody, Abnova™

Catalog No. 89117784 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-117-784 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-117-784 Supplier Abnova Corporation Supplier No. PAB15719
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against partial recombinant Pde4dip .

Sequence: TMQSMESLLRARGLEVEQLTATCQNLQWLKEELETKFGHWQKEQESIIQQLQTSLHDRNKEVEDLSATLLCKLGPGQSEVAEELCQRLQRKERMLQDLLSDRNKQAVEHEMEIQGLLQSMGTREQERQAAAEKMVQAFMERNSELQALRQYLGGKELMTSSQTFISNQPAGVTSIGPHHGEQTDQGSMQMPSRDDSTSLTAREEASIPRSTLGDSDTVAGLEKELSNAKEELELMAKKERESQMELSALQSMMAMQEEELQVQAADLESLTRNVQIKEDLIKDLQMQ

Specifications

Antigen Pde4dip
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against partial recombinant Pde4dip .
Dilution Western Blot (1:1000) The optimal working dilution should be determined by the end user.
Formulation In PBS (50% glycerol, 0.02% sodium azide)
Gene Pde4dip
Gene Alias 4732458A06Rik/9430063L05Rik/C87016/D130016K21Rik/D3Bwg1078e/KIAA0477/MGC176146/Usmg4/mKIAA0454
Gene Symbols Pde4dip
Host Species Rabbit
Immunogen Recombinant GST fusion protein corresponding to 287 mouse Pde4dip .
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 83679
Test Specificity Specific to recombinant protein GX0271. This antibody detects mPDE4DIP protein.
Target Species Mouse
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.