Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PDHA1, Rabbit, HRP, Polyclonal Antibody, Abnova™

Catalog No. 89117568 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-117-568 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-117-568 Supplier Abnova Corporation Supplier No. PAB15338
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against synthetic phosphopeptide of PDHA1.

The pyruvate dehydrogenase complex is a nuclear-encoded mitochondrial matrix multienzyme complex that provides the primary link between glycolysis and the tricarboxylic acid (TCA) cycle by catalyzing the irreversible conversion of pyruvate into acetyl-CoA. The PDH complex is composed of multiple copies of 3 enzymes: E1 (PDHA1); dihydrolipoyl transacetylase (DLAT; MIM 608770) (E2; EC 2.3.1.12); and dihydrolipoyl dehydrogenase (DLD; MIM 238331) (E3; EC 1.8.1.4). The E1 enzyme is a heterotetramer of 2 alpha and 2 beta subunits. The E1-alpha subunit contains the E1 active site and plays a key role in the function of the PDH complex (Brown et al., 1994 [PubMed 7853374]).[supplied by OMIM

Specifications

Antigen PDHA1
Applications Western Blot
Classification Polyclonal
Conjugate HRP
Description Rabbit polyclonal antibody raised against synthetic phosphopeptide of PDHA1.
Dilution Western Blot (1:2500-1:5000) The optimal working dilution should be determined by the end user.
Formulation In PBS
Gene PDHA1
Gene Alias PDHA/PDHCE1A/PHE1A
Gene Symbols PDHA1
Host Species Rabbit
Immunogen Synthetic phosphopeptide corresponding to residues surrounding S293 human PDHA1.
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5160
Test Specificity This is specific to the phosphorylated Serine 293 form of the PDHE1 alpha protein. This antibody is useful for Western blot, where a band is seen ∼43 KDa.
Target Species Human
Content And Storage Store at 4°C. Do not freeze.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.