Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PDIA2, Rabbit, Polyclonal Antibody, Abnova™

Catalog No. 89115930 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-115-930 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-115-930 Supplier Abnova Corporation Supplier No. PAB12587
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against full-length native PDIA2.

Protein disulfide isomerases (EC 5.3.4.1), such as PDIP, are endoplasmic reticulum (ER) resident proteins that catalyze protein folding and thiol-disulfide interchange reactions (Desilva et al., 1996 [PubMed 8561901]).[supplied by OMIM

Specifications

Antigen PDIA2
Applications Immunocytochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (PFA fixed), Immunoprecipitation, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against full length native PDIA2.
Dilution Immunocytochemistry (1:5000-1:20000) Immunohistochemistry (Frozen sections) (1:100-1:1000) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (1:100-1:500) Immunoprecipitation (1-3 ug) Western Blot (1:10000-1:20000) The op
Formulation In antiserum
Gene PDIA2
Gene Alias PDA2/PDI/PDIP/PDIR
Gene Symbols PDIA2
Host Species Rabbit
Immunogen Native purified PDIA2.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 64714
Target Species Bovine, Human, Mouse, Primate, Rat
Content And Storage Store at -20°C or -80°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.