Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PDK1 Rabbit anti-Human, DyLight 549, Polyclonal Antibody, Abnova™

Catalog No. 89117562 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-117-562 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-117-562 Supplier Abnova Corporation Supplier No. PAB15332
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against synthetic peptide of PDK1.

Pyruvate dehydrogenase (PDH) is a mitochondrial multienzyme complex that catalyzes the oxidative decarboxylation of pyruvate and is one of the major enzymes responsible for the regulation of homeostasis of carbohydrate fuels in mammals. The enzymatic activity is regulated by a phosphorylation/dephosphorylation cycle. Phosphorylation of PDH by a specific pyruvate dehydrogenase kinase (PDK) results in inactivation. [provided by RefSeq

Specifications

Antigen PDK1
Applications Western Blot
Classification Polyclonal
Conjugate DyLight 549
Description Rabbit polyclonal antibody raised against synthetic peptide of PDK1.
Dilution Western Blot (2 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In 50mM sodium borate
Gene PDK1
Gene Symbols PDK1
Host Species Rabbit
Immunogen A synthetic peptide corresponding to amino acids 350-436 of human PDK1.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5163
Test Specificity This antibody reacts with human. No other species have been tested. The immunogen used for this antibody production has 100% homology with mouse and rat PDK1 protein. This is specific to PDK1. By Western blot a band is seen at ∼49 KDa, representing PDK1 protein. There are also two higher and one lower non-specific molecular weight bands.
Target Species Human
Content And Storage Store at 4°C. Do not freeze.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.