Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PDPK1 Rabbit anti-Human, Mouse, Rat, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of PDPK1.
Supplier: Abnova Corporation PAB7885
Specifications
| PDPK1 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of PDPK1. | |
| In PBS (50% glycerol, 1mg/mL BSA, 0.05% sodium azide) | |
| MGC20087/MGC35290/PDK1/PRO0461 | |
| Rabbit | |
| 100 μL | |
| Primary | |
| This sequence is highly conserved in rat and mouse PDK1. | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
| ELISA, Western Blot | |
| Unconjugated | |
| ELISA (1:2000) Western Blot (1:1000) The optimal working dilution should be determined by the end user. | |
| PDPK1 | |
| PDPK1 | |
| A synthetic peptide corresponding to N-terminus of human PDPK1. | |
| RUO | |
| 5170 | |
| Human, Mouse, Rat | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction