Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PDZK1, Rabbit, Polyclonal Antibody, Abnova™

Catalog No. 89117643 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-117-643 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-117-643 Supplier Abnova Corporation Supplier No. PAB15564
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against synthetic peptide of PDZK1.

Specifications

Antigen PDZK1
Applications Immunocytochemistry, Immunofluorescence, Immunohistochemistry (PFA fixed), Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against synthetic peptide of PDZK1.
Dilution Immunocytochemistry (1:200) Immunofluorescence (1:200) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (1:400) Western Blot (1:1000) The optimal working dilution should be determined by the end user.
Formulation In antiserum (0.05% sodium azide)
Gene PDZK1
Gene Alias CAP70/CLAMP/NHERF3/PDZD1
Gene Symbols PDZK1
Host Species Rabbit
Immunogen A synthetic peptide corresponding to C-terminus of Human PDZK1.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5174
Test Specificity This is specific to PDZK1. Does not react with rat. A single band of ∼70 KDa is detected by Western blot (SDS-PAGE). No other application has been tested.
Target Species Bovine, Human, Mouse
Content And Storage Store at -20°C or -80°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.