Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PECAM1, Rabbit, Clone: SP164, Abnova™

Catalog No. 89109257 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-109-257 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-109-257 Supplier Abnova Corporation Supplier No. MAB11043

Rabbit monoclonal antibody raised against partial recombinant PECAM1.

Specifications

Antigen PECAM1
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone SP164
Conjugate Unconjugated
Description Rabbit monoclonal antibody raised against partial recombinant PECAM1.
Dilution Immunohistochemistry (1:50) The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.6 (1% BSA, 0.09% sodium azide)
Gene Pecam1
Gene Alias C85791/Cd31/MGC102160/PECAM-1/Pecam
Gene Symbols Pecam1
Host Species Rabbit
Immunogen A synthetic peptide corresponding to C-terminus of mouse PECAM1.
Purification Method Protein A/G purification
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 18613
Target Species Human
Content And Storage Store at 4°C. Do not freeze.
Form Liquid
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.