Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PGBD3, Rabbit, Polyclonal Antibody, Abnova™

Catalog No. 89123488 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-123-488 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-123-488 Supplier Abnova Corporation Supplier No. PAB21786

Rabbit polyclonal antibody raised against recombinant PGBD3.

The piggyBac family of proteins, found in diverse animals, are transposases related to the transposase of the canonical piggyBac transposon from the moth, Trichoplusia ni. This family also includes genes in several genomes, including human, that appear to have been derived from the piggyBac transposons. This gene belongs to the subfamily of piggyBac transposable element derived (PGBD) genes. The PGBD proteins appear to be novel, with no obvious relationship to other transposases, or other known protein families. This gene overlaps with the ERCC6 gene on chromosome 10, and pseudogenes of this locus have been found on chromosomes 4, 5 and 12. [provided by RefSeq

Sequence: VYNQFMGGVDRADENIDKYRASIRGKKWYSSPLLFCFELVLQNAWQLHKTYDEKPVDFLEFRRRVVCHYLETHGHPPEPGQKGRPQKRNIDSRYDGINHVIVKQGKQTRCAECHKNTTFRCEKCDVALHVKCSVEY

Specifications

Antigen PGBD3
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description piggyBac transposable element derived 3
Formulation In PBS, pH 7.5 (40% glycerol, 0.02% sodium azide)
Gene PGBD3
Gene Alias FLJ90201
Gene Symbols PGBD3
Host Species Rabbit
Immunogen Recombinant protein corresponding to amino acids of human PGBD3.
Purification Method Affinity Purified
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 267004
Target Species Human
Content And Storage Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.