Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PGR, Rabbit, Clone: A21-W, Abnova™

Catalog No. 89113779 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
500 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-113-779 500 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-113-779 Supplier Abnova Corporation Supplier No. MAB9785
Add to Cart
Add to Cart

Rabbit monoclonal antibody raised against synthetic peptide of PGR.

This gene encodes a member of the steroid receptor superfamily. The encoded protein mediates the physiological effects of progesterone, which plays a central role in reproductive events associated with the establishment and maintenance of pregnancy. This gene uses two distinct promotors and translation start sites in the first exon to produce two isoforms, A and B. The two isoforms are identical except for the additional 165 amino acids found in the N-terminus of isoform A only, and mediate their own response genes and physiologic effects with little overlap. The location of transcription initiation for isoform B has not been clearly determined. [provided by RefSeq

Specifications

Antigen PGR
Applications Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone A21-W
Conjugate Unconjugated
Description Rabbit monoclonal antibody raised against synthetic peptide of PGR.
Dilution Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (1:100-1:200) The optimal working dilution should be determined by the end user.
Formulation In 20mM Tris-HCl, pH 8.0 (20mg/mL BSA, 0.05% sodium azide)
Gene PGR
Gene Accession No. P06401
Gene Alias NR3C3/PR
Gene Symbols PGR
Host Species Rabbit
Immunogen A synthetic peptide corresponding to N-terminus of human PGR.
Quantity 500 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5241
Target Species Human, Mouse
Content And Storage Store at 4°C.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.