Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Rabbit anti-PHLPP2, Polyclonal,

Catalog No. ABS1017MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ABS1017MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ABS1017MI Supplier MilliporeSigma™ Supplier No. ABS1017
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Specifically detects PHLPP2 in Human samples, and it is validated for Immunocytochemistry, Western Blotting

PH Domain And Leucine Rich Repeat Protein Phosphatase 2 is encoded by the PHLPP2 gene. It is known to catalyze the dephosphorylation of pSer473 of AKT1 and pSer660 of PRKCbeta-II, as well as pSer657 of PRKCalpha.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PHLPP2
Applications Immunocytochemistry, Western Blot
Classification Polyclonal
Formulation Purified rabbit polyclonal in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide.
Gene Accession No. Q6ZVD8
Gene Symbols PHLPP2; KIAA0931; PHLPPL
Host Species Rabbit
Immunogen KLH-conjugated linear peptide corresponding within the PP2C-like domain of human PHLPP2.
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Signaling
Primary or Secondary Primary
Gene ID (Entrez) NP_055835
Target Species Human
Content And Storage Stable for one year at 2°C to 8°C from date of receipt.
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.