Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Rabbit anti-phospho-AS160 (Thr642), Polyclonal,

Catalog No. ABS271MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ABS271MI 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ABS271MI Supplier MilliporeSigma™ Supplier No. ABS271
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Specifically detects phospho-AS160 (Thr642) in Human, Mouse samples, and it is validated for Western Blotting

AS160 is a widely expressed RAB-GTPase activating protein which plays a role in insulin action. AS160 is phosphorylated by AKT, AMPK, RSK, and SGK1. Upon phosphorylation, AS160 changes its location from microsomes to the cytosol upon insulin treatment. AS160 acts as key link between insulin signaling and GLUT4 trafficking of glucose. Phosphorylation of AS160 is essential for AS160 action and AS160 has multiple phosphorylation sites. Phosphorylation of AS160 on both AS160Thr642 and AS160Ser588 is important for insulin-stimulated glucose transport. Phosphorylation of these sites is predicted to inhibit AS160's activation of Rab-GTPase proteins and favor greater exocytosis of GLUT4 vesicles, leading ultimately to greater glucose transport. Research indicates that impairment of phosphorylation of AS160 at Thr642 is associated with various aspects of type 2 diabetes.
TRUSTED_SUSTAINABILITY

Specifications

Antigen phospho-AS160 (Thr642)
Applications Western Blot
Classification Polyclonal
Formulation Purified rabbit polyclonal in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide.
Gene Accession No. O60343
Gene Symbols TBC1D4; AS160; KIAA0603
Host Species Rabbit
Immunogen KLH-conjugated linear peptide corresponding to human phospho-AS160 (Thr642).
Purification Method Affinity Purified
Quantity 200 μL
Regulatory Status RUO
Research Discipline Signaling
Primary or Secondary Primary
Gene ID (Entrez) NP_055647
Target Species Human, Mouse
Content And Storage Stable for one year at 2°C to 8°C from date of receipt.
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.