Learn More
MilliporeSigma™ Rabbit anti-phospho-CREB (Ser133), Alexa Fluor 647, Polyclonal,
Shop All MilliporeSigma ProductsDescription
Specifically detects phospho-CREB (Ser133) in Human, Mouse, Rat samples, and it is validated for Immunofluorescence
Specifications
Specifications
| Antigen | phospho-CREB (Ser133) |
| Applications | Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Alexa Fluor 647 |
| Formulation | Purified rabbit polyclonal antibody conjugate in PBS with 15mg/mL BSA and 0.1% sodium azide. |
| Gene Accession No. | P16220 |
| Gene Symbols | CREB1 |
| Host Species | Rabbit |
| Immunogen | KLH-conjugated synthetic peptide corresponding to the amino acids including and encompassing phosphorylated Ser133 of CREB-A. |
| Purification Method | Affinity Purified |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.