Learn More
MilliporeSigma™ Rabbit anti-phospho-Neph1 (Tyr637/638), Polyclonal,
Shop All MilliporeSigma ProductsDescription
Specifically detects phospho-Neph1 (Tyr637/638) in Human samples, and it is validated for Western Blotting
Specifications
Specifications
| Antigen | phospho-Neph1 (Tyr637/638) |
| Applications | Western Blot |
| Classification | Polyclonal |
| Formulation | Purified rabbit polyclonal antibody in buffer containing PBS/glycine/5% BSA, 0.02% Sodium Azide and 30% glycerol. |
| Gene Accession No. | Q80W68 |
| Gene Symbols | Kirrel; Kirrel1; Neph1 |
| Host Species | Rabbit |
| Immunogen | Linear peptide corresponding to the cytoplasmic domain of mouse phospho-Neph1 (Tyr637/638). |
| Purification Method | Affinity Purified |
| Quantity | 100 μL |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.