Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Rabbit anti-phospho- NFkB p52, (Ser222), Polyclonal,

Catalog No. ABS1515MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ABS1515MI 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ABS1515MI Supplier MilliporeSigma™ Supplier No. ABS1515
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Specifically detects phospho- NFkB p52, (Ser222) in Human samples, and it is validated for Western Blotting

The p52 NF-kB subunit is induced through activation of the alternative (noncanonical) NF-kB pathway in response to a range of stimuli such as lymphotoxin-b, BAFF and CD40, as well as DNA damage. The p52 formation results from proteolytic processing of its precursor protein p100 as a consequence of IKKalpha-mediated phosphorylation. The p52 is most commonly found as a heterodimer with RelB, although other dimer combinations have been identified by ChIP analysis. Ser222 of NF-kappa B p52 is phosphorylated by the kinase GSK3beta. Phosphorylation at S222 appears to regulate not only p52 partner dimerization but also the p52 dependent activation or repression of Skp2 gene expression.
TRUSTED_SUSTAINABILITY

Specifications

Antigen phospho- NFkB p52, (Ser222)
Applications Western Blot
Classification Polyclonal
Formulation Purified rabbit polyclonal in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide.
Gene Accession No. Q00653
Gene Symbols NFKB2; LYT10
Host Species Rabbit
Immunogen KLH-conjugated linear peptide corresponding to the RHD domain of human phospho NFkB p52, (Ser222).
Purification Method Affinity Purified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Signaling
Primary or Secondary Primary
Gene ID (Entrez) NP_002493
Target Species Human
Content And Storage Stable for one year at 2°C to 8°C from date of receipt.
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.