Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Rabbit anti-phospho- PSD95 (Thr19), Polyclonal,

Catalog No. ABN998MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ABN998MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ABN998MI Supplier MilliporeSigma™ Supplier No. ABN998
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Specifically detects phospho- PSD95 (Thr19) in Human samples, and it is validated for Immunohistochemistry (Paraffin), Western Blotting

PSD95 (postsynaptic density95) represent a class of anchoring proteins which serve to localize various neuronal ion channels to post-synaptic densities. PSD95 and related proteins contain PDZ domains, which interact with NMDA receptors and voltage-gated K+ channels via a ET/SXV motif in the cytoplasmic carboxyl terminus of the receptor channels.
TRUSTED_SUSTAINABILITY

Specifications

Antigen phospho- PSD95 (Thr19)
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Formulation Purified rabbit polyclonal in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide.
Gene Accession No. P78352
Gene Symbols DLG4; PSD95
Host Species Rabbit
Immunogen KLH-conjugated linear peptide corresponding to human phospho PSD95 (Thr19) near the N-Terminus.
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) NP_001122299
Target Species Human
Content And Storage Stable for one year at 2°C to 8°C from date of receipt.
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.