Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Rabbit anti-phospho-Rac1/cdc42 (Ser71), Polyclonal,

Catalog No. 07896IMI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
07-896-IMI 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 07-896-IMI Supplier MilliporeSigma™ Supplier No. 07896I
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Specifically detects phospho-Rac1/cdc42 (Ser71) in Human samples, and it is validated for Immunoprecipitation, Western Blotting

Target of rapamycin complex 2 subunit MAPKAP1 (UniProt Q9BPZ7; also known as MEKK2-interacting protein 1, Mitogen-activated protein kinase 2-associated protein 1, mSIN1, Ras inhibitor MGC2745, SAPK-interacting protein 1, Stress-activated protein kinase-interacting 1, Stress-activated map kinase-interacting protein 1,, TORC2 subunit MAPKAP1) is encoded by the MAPKAP1 (also known as JC310, MIP1 SIN1, SIN1b, SIN1g) gene (Gene ID 79109) in human. mSIN1 is an essential component of the Torc2 complex. mTORC2 is comprised of mTOR, a large Ser/Thr protein kinase, along with mSIN1, GbL (mLST8), Protor1, Protor2, and Rictor. The mTORC2 complex is activated primarily downstream of PI3 Kinase and is known to affect cell proliferation and survival primarily by phosphorylating Akt on Ser473. Additionally, the mTORC2 complex is known to affect cytoskeletal organization and cell migration by exerting its effects through Rac, Rho, and PKC. mSIN1 is also known to directly interact with Ras, SPAKs, and JNK.
TRUSTED_SUSTAINABILITY

Specifications

Antigen phospho-Rac1/cdc42 (Ser71)
Applications Immunoprecipitation, Western Blot
Classification Polyclonal
Formulation Purified rabbit polyclonal antibody in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide.
Gene Accession No. P63000
Gene Symbols RAC1; TC25; MIG5
Host Species Rabbit
Immunogen KLH-conjugated linear peptide corresponding to human phospho-Rac1/cdc42 (Ser71) near the N-Terminus.
Purification Method Affinity Purified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Signaling
Primary or Secondary Primary
Gene ID (Entrez) NP_008839
Target Species Human
Content And Storage Stable for one year at 2°C to 8°C from date of receipt.
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.