Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma™ Rabbit anti-phospho-Raptor (Ser696), Polyclonal,
Rabbit Polyclonal Antibody
Supplier: MilliporeSigma™ ABS544
Description
Specifically detects phospho-Raptor (Ser696) in Human samples, and it is validated for Western Blotting
Raptor is a subunit of mTORC1, which regulates cell growth and survival in response to nutrient and hormonal signals. mTORC1 is activated in response to growth factors or amino-acids. Within mTORC1, Raptor positively regulates mTORC1 activity and functions as a scaffold for recruiting mTORC1 substrates. Recognizes Raptor that is phosphorylated at serine 696.
Specifications
| phospho-Raptor (Ser696) | |
| Polyclonal | |
| Q8N122 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human |
| Western Blot | |
| Purified rabbit polyclonal in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide. | |
| RPTOR; KIAA1303; RAPTOR | |
| KLH-conjugated linear peptide corresponding to human phospho-Raptor (Ser696). | |
| 200 μL | |
| Signaling | |
| NP_065812 | |
| Stable for one year at 2°C to 8°C from date of receipt. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction