Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Rabbit anti-phospho- SIRT1 (Ser682), Polyclonal,

Catalog No. ABE1392MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ABE1392MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ABE1392MI Supplier MilliporeSigma™ Supplier No. ABE1392
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Specifically detects phospho- SIRT1 (Ser682) in Human samples, and it is validated for Western Blotting

NAD-dependent protein deacetylase sirtuin-1 (UniProt Q96EB6; also known as hSIR2, hSIRT1, NAD-dependent deacetylase sirtuin-1, Regulatory protein SIR2 homolog 1, Sir2-like 1, SIR2-like protein 1, SIR2alpha, Sirtuin type 1) is encoded by the SIRT1 (also known as SIR2L1) gene (Gene ID 23411) in human. Sirtuins constitute a family of NAD+-dependent deacetylases found in nearly all organisms studied. Sirtuins have been implicated in the regulation of multiple processes, including aging, transcription, apoptosis, and stress resistance. Full-length SirT1 (FLSirT1) plays an important role in human cartilage homeostasis and chondrocyte survival by mediating the expression of major cartilage anabolic components, collagen 2(I) and aggrecan. The generation of the transcriptionally inactive 75-kDa fragment (75SirT1) via site-specific cleavage of FLSirT1 at amino acid 533 by cathepsin B is an anti-apoptotic mechanism to promote human osteoarthritic (OA) chondrocytes survival following exposure to proinflammatory cytokines. The 5SirT1 fragment is shown to associate with cytochrome c and likely blocks downstream apoptosome assembly. Phosphorylation plays an important role in modulating SirT1 activity. Multiple kinases and their corresponding SirT1 phosphorylation sites have been identified, including pT344 targeted by AMPK, pT530 and pS540 targeted by Cdk1/Cyclin B, pS46 (mouse) or pS47 (human) targeted by JNK1, pT522 targeted by DYRK1A & DYRK3, as well as pS27 and pS47 targeted by CaMKKβ.
TRUSTED_SUSTAINABILITY

Specifications

Antigen phospho- SIRT1 (Ser682)
Applications Western Blot
Classification Polyclonal
Formulation Purified rabbit polyclonal antibody in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide.
Gene Accession No. Q96EB6
Gene Symbols SIRT1; SIR2L1
Host Species Rabbit
Immunogen KLH-conjugated linear peptide corresponding to human phospho SIRT1 (Ser682) near the C-terminus.
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Epigenetics and Nuclear Function
Primary or Secondary Primary
Gene ID (Entrez) NP_036370
Target Species Human
Content And Storage Stable for one year at 2°C to 8°C from date of receipt.
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.