Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Rabbit anti-phospho- SMAD2 (Ser465/467), Polyclonal,

Catalog No. AB3849I Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AB3849I 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AB3849I Supplier MilliporeSigma™ Supplier No. AB3849I
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Specifically detects phospho- SMAD2 (Ser465/467) in Human samples, and it is validated for Western Blotting

Mothers against decapentaplegic homolog 2 (UniProt Q15796; also known as hMAD-2, hSMAD2, JV18-1, MAD homolog 2, Mad-related protein 2, Mothers against DPP homolog 2, SMAD 2, SMAD family member 2, Smad2) is encoded by the SMAD2 (also known as MADH2, MADR2) gene (Gene ID 4087) in human. Smad2 is a receptor-regulated intracellular signal transducer and transcriptional modulator activated by transforming growth factor TGF-beta and activin type 1 receptor kinases. In response to TGF-beta signal, smad2 is recruited to the activated TGF-beta receptor through its interaction with the SMAD anchor for receptor activation (SARA) protein, where it is phosphorylated by the TGF-beta receptor. The phosphorylation results in its dissociation from SARA and association with another family member smad4. The smad2-smad4 complex then translocates into the nucleus, where it binds to target promoters and forms a transcription repressor complex with other cofactors. Smad2 can also be phosphorylated by activin type 1 receptor kinase, and thereby transduce activin-dependent signaling events.
TRUSTED_SUSTAINABILITY

Specifications

Antigen phospho- SMAD2 (Ser465/467)
Applications Western Blot
Classification Polyclonal
Formulation Purified rabbit polyclonal antibody in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide.
Gene Accession No. Q15796
Gene Symbols SMAD2; MADH2; MADR2
Host Species Rabbit
Immunogen KLH-conjugated, synthetic phospho-peptide corresponding to residues surrounding Ser465/467 of human Smad2.
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Epigenetics and Nuclear Function
Primary or Secondary Primary
Gene ID (Entrez) NP_005892
Target Species Human
Content And Storage Stable for one year at 2°C to 8°C from date of receipt.
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.