Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Rabbit anti-phospho- Synaptotagmin-12 (Ser97), Polyclonal,

Catalog No. ABN997MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ABN997MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ABN997MI Supplier MilliporeSigma™ Supplier No. ABN997
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Specifically detects phospho- Synaptotagmin-12 (Ser97) in Mouse samples, and it is validated for Western Blotting

Synaptotagmin-12 (also called Synaptotagmin XII or SytXII) is a member of the synaptotagmin family of transmembrane transporters. Synaptotagmin-12 is thought to be involved in Ca2+-dependent exocytosis of secretory vesicles by functioning as a Ca2+ sensor during vesicular trafficking and exocytosis process.
TRUSTED_SUSTAINABILITY

Specifications

Antigen phospho- Synaptotagmin-12 (Ser97)
Applications Western Blot
Classification Polyclonal
Formulation Purified rabbit polyclonal in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide.
Gene Accession No. Q8IV01
Gene Symbols SYT12
Host Species Rabbit
Immunogen KLH-conjugated linear peptide corresponding to human phospho Synaptotagmin-12 (Ser97).
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) NP_808878
Target Species Mouse
Content And Storage Stable for one year at 2°C to 8°C from date of receipt.
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.