Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Rabbit anti-phospho-TrkB (Tyr816), Polyclonal,

Catalog No. ABN1381MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ABN1381MI 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ABN1381MI Supplier MilliporeSigma™ Supplier No. ABN1381
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Specifically detects phospho-TrkB (Tyr816) in Mouse, Rat samples, and it is validated for Immunocytochemistry, Immunohistochemistry, Immunoprecipitation, Western Blotting

BDNF/NT-3 growth factors receptor (EC 2.7.10.1; UniProt Q63604; also known as GP145-TrkB/GP95-TrkB, Trk-B, TrkB tyrosine kinase, Neurotrophic tyrosine receptor kinase type 2, Neural receptor protein-tyrosine kinase) is encoded by the Ntrk2 gene (also known as RATTRKB1, Trkb, TRKB1) in rat (Gene ID 25054). Neurotrophins mediate a wide range of neuronal functions, including survival, differentiation, growth, and death, via two classes of transmembrane receptors, the Trk receptor tyrosine kinase and the p75 neurotrophin receptor. While nerve growth factor (NGF), brain-derived neurotrophic factor (BDNF), neurotrophin-3 (NT-3), and NT-4 all bind the p75 receptor, TrkA is selective for NGF, TrkB is selective for BDNF and NT-4, and TrkC is selective for NT-3. TrkA and TrkB receptors can also be activated by ligands of the G-protein-coupled receptor (GPCRs), including the nucleoside adenosine or adenosine agonists, such as CGS 21680 and the pituitary adenylate cyclase-activating polypeptide (PACAP). In addition to transactivation via GPCRs and glucocorticoids are also reported to promote TrkA/B/C activation due to signaling cross-talk between Trk and GC receptor (GR).
TRUSTED_SUSTAINABILITY

Specifications

Antigen phospho-TrkB (Tyr816)
Applications Immunocytochemistry, Immunohistochemistry, Immunoprecipitation, Western Blot
Classification Polyclonal
Formulation Purified rabbit polyclonal antibody in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide.
Gene Accession No. Q63604
Gene Symbols Ntrk2; Trkb
Host Species Rabbit
Immunogen Linear peptide corresponding to rat TrkB sequence near the C-terminus with phosphorylated Tyr816.
Purification Method Affinity Purified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) NP_036863
Target Species Mouse, Rat
Content And Storage Stable for one year at 2°C to 8°C from date of receipt.
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.