Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

phosphoserine, Rabbit, Polyclonal Antibody, Abnova™

Catalog No. 89138206 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-138-206 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-138-206 Supplier Abnova Corporation Supplier No. PAB16619
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against phosphoserine.

Specifications

Antigen phosphoserine
Applications ELISA, Immunohistochemistry, Immunoprecipitation, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against phosphoserine.
Dilution ELISA (0.5-4 ug/mL) Western Blot (2-4 ug/mL) Immunoprecipitation (10 ug/mg lysate) The optimal working dilution should be determined by the end user.
Formulation In PBS (50% glycerol, 0.09% sodium azide)
Host Species Rabbit
Immunogen Phosphoserine conjugated with KLH.
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity Recognizes proteins phosphorylated on serine residues. Does not cross-react with phosphotyrosine. Species independent of cross reactivity.
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.