Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Phosphoserine Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against Phosphoserine.
Supplier: Abnova Corporation PAB10351
Specifications
Phosphoserine | |
Polyclonal | |
Rabbit polyclonal antibody raised against Phosphoserine. | |
In 20mM KH2PO4, 150mM NaCl, pH 7.2 | |
Phosphoserine conjugated with KLH. | |
RUO | |
Reactivity is specific to phosphoserine and minimal cross-reactivity is observed against phosphotyrosine or phosphothreonine. | |
Liquid |
ELISA, Immunohistochemistry, Immunoprecipitation, Western Blot | |
Unconjugated | |
ELISA (1:1000-1:2500) Western Blot (1:500-1:1000) Immunoprecipitation (10 ug/mg protein sample) The optimal working dilution should be determined by the end user. | |
Rabbit | |
100 μg | |
Primary | |
Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction