Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phosphotyrosine Rabbit, Polyclonal Antibody, Abnova™

Catalog No. 89136965 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-136-965 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-136-965 Supplier Abnova Corporation Supplier No. PAB10353
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against Phosphotyrosine.

Specifications

Antigen Phosphotyrosine
Applications ELISA, Immunohistochemistry, Immunoprecipitation, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against Phosphotyrosine.
Dilution ELISA (1:1000-1:5000) Western Blot (1:500) Immunoprecipitation (10.0 ug/ 500 ug sample) Immunohistochemistry (1:50) The optimal working dilution should be determined by the end user.
Formulation In 20mM KH2PO4, 150mM NaCl, pH 7.2 (0.01% sodium azide)
Host Species Rabbit
Immunogen Phosphotyrosine conjugated with KLH.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity Reactivity is specific to phosphotyrosine and minimal cross reactivity is observed against phosphothreonine or phosphoserine.
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.