Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

phototropin 2, Rabbit, Polyclonal Antibody, Abnova™

Catalog No. 89133717 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-133-717 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-133-717 Supplier Abnova Corporation Supplier No. PAB8516
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against synthetic peptide of phototropin 2.

Sequence: NDAESLSERRSC

Specifications

Antigen phototropin 2
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against synthetic peptide of phototropin 2.
Dilution Western Blot (2-5 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In PBS (0.1% proclin, 2.0% Block Ace)
Gene PHOT2
Gene Alias K21L19.6/K21L19_6/NON PHOTOTROPIC HYPOCOTYL 1-LIKE/NPL1
Gene Symbols PHOT2
Host Species Rabbit
Immunogen A synthetic peptide corresponding to amino acids 12-22 at N-terminus of arabidopsis phototropin 2
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 835926
Target Species Thale cress
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.