Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PIGY Rabbit anti-Human, Mouse, Polyclonal Antibody, Abnova™

Catalog No. 89118470 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-118-470 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-118-470 Supplier Abnova Corporation Supplier No. PAB16599
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against synthetic peptide of PIGY.

This gene encodes two proteins, one of which is part of the GPI-N-acetylglucosaminyltransferase (GIP-GnT) complex which initiates the biosynthesis of glycosylphosphatidylinositol (GPI). GPI is synthesized in the endoplasmic reticulum and serves as an anchor for many surface proteins. Proteins containing GPI anchors can have an important role in cell-cell interactions. Two open reading frames have been found in the single transcript that has been identified for this gene. The downstream open reading frame encodes the GPI-GnT complex protein while the upstream open reading frame encodes a protein with unknown function. [provided by RefSeq

Specifications

Antigen PIGY
Applications ELISA, Immunohistochemistry, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against synthetic peptide of PIGY.
Dilution Western Blot (1-2 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In PBS (0.02% sodium azide)
Gene PIGY
Gene Alias MGC14156/PIG-Y
Gene Symbols PIGY
Host Species Rabbit
Immunogen A synthetic peptide corresponding to C-terminus 16 amino acids of human PIGY.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84992
Target Species Human, Mouse
Content And Storage Store at 4°C for three months. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.