Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PIK3C3 Rabbit anti-Human, Polyclonal Antibody, Abnova™

Catalog No. 89119623 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
200 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-119-623 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-119-623 Supplier Abnova Corporation Supplier No. PAB18040

Rabbit polyclonal antibody raised against synthetic peptide of PIK3C3.

Specifications

Antigen PIK3C3
Applications ELISA
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against synthetic peptide of PIK3C3.
Dilution Recommend working dilution at 1:32000. The optimal working dilution should be determined by the end user.
Formulation In PBS.
Gene PIK3C3
Gene Alias MGC61518/Vps34
Gene Symbols PIK3C3
Host Species Rabbit
Immunogen A synthetic peptide corresponding to human PIK3C3.
Quantity 200 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5289
Test Specificity The rabbit anti-human PIK3C3 antibody detects specifically synthetic antigen derived from human PIK3C3 at various concentrations. Reactivity with cellular proteins is not tested yet. Cross reactivity with other species are expected from sequence similarity.
Target Species Human
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.