Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PIK3CA, Rabbit, Clone: SP139, Abnova™

Catalog No. 89109259 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-109-259 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-109-259 Supplier Abnova Corporation Supplier No. MAB11045

Rabbit monoclonal antibody raised against synthetic peptide of PIK3CA.

Phosphatidylinositol 3-kinase is composed of an 85kDa regulatory subunit and a 110kDa catalytic subunit. The protein encoded by this gene represents the catalytic subunit, which uses ATP to phosphorylate PtdIns, PtdIns4P and PtdIns(4,5)P2. This gene has been found to be oncogenic and has been implicated in cervical cancers. [provided by RefSeq

Specifications

Antigen PIK3CA
Applications Immunohistochemistry (PFA fixed)
Classification Monoclonal
Clone SP139
Conjugate Unconjugated
Description Rabbit monoclonal antibody raised against synthetic peptide of PIK3CA.
Dilution Immunohistochemistry (1:100) The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.6 (1% BSA, 0.09% sodium azide)
Gene PIK3CA
Gene Alias MGC142161/MGC142163/PI3K/p110-alpha
Gene Symbols PIK3CA
Host Species Rabbit
Immunogen A synthetic peptide corresponding to internal region of human PIK3CA.
Purification Method Protein A/G purification
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5290
Target Species Human
Content And Storage Store at 4°C. Do not freeze.
Form Liquid
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.