Learn More
PIK3CD Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of PIK3CD.
Supplier: Abnova Corporation PAB3214
Description
Phosphoinositide 3-kinases (PI3Ks) phosphorylate the 3-prime OH position of the inositol ring of inositol lipids. See MIM 602838. The class I PI3Ks display a broad phosphoinositide lipid substrate specificity and include p110-alpha (MIM 171834), p110-beta (MIM 602925), and p110-gamma (MIM 601232). p110-alpha and p110-beta interact with SH2/SH3-domain-containing p85 adaptor proteins (see MIM 171833) and with GTP-bound Ras.[supplied by OMIM
Specifications
PIK3CD | |
Polyclonal | |
Rabbit polyclonal antibody raised against synthetic peptide of PIK3CD. | |
In PBS (0.09% sodium azide) | |
p110D | |
Rabbit | |
Protein G purification | |
RUO | |
5293 | |
Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Immunofluorescence, Immunohistochemistry (PFA fixed), Western Blot | |
Unconjugated | |
Immunofluorescence (1:10-50) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (1:50-100) Western Blot (1:1000) The optimal working dilution should be determined by the end user. | |
PIK3CD | |
PIK3CD | |
A synthetic peptide (conjugated with KLH) corresponding to C-terminus of human PIK3CD. | |
400 μL | |
Primary | |
Human | |
Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only