Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PIK3R1/PIK3R3 (phospho Y467/Y199), Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic phosphopeptide of PIK3R1/PIK3R3.
Supplier: Abnova Corporation PAB24721
Specifications
PIK3R1/PIK3R3 | |
Polyclonal | |
Rabbit polyclonal antibody raised against synthetic phosphopeptide of PIK3R1/PIK3R3. | |
In PBS, pH 7.2 (0.09% sodium azide) | |
PIK3R1 | |
Synthetic phosphopeptide corresponding to residues surrounding Y467/Y199 of PIK3R1/PIK3R3. | |
100 μL | |
Primary | |
PIK3R1/PIK3R3 (phospho Y467/Y199) polyclonal antibody detects endogenous levels of PIK3R1/PIK3R3 protein. | |
Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Western Blot | |
Unconjugated | |
Western Blot (1:500-1:1000) The optimal working dilution should be determined by the end user. | |
GRB1/p85/p85-ALPHA | |
Rabbit | |
Antigen affinity purification | |
RUO | |
5295/8503 | |
Human | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction