Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PIN1 Rabbit anti-Bovine, Dog, Human, Monkey, Mouse, Rat, Polyclonal Antibody, Abnova™

Catalog No. 89114953 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-114-953 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-114-953 Supplier Abnova Corporation Supplier No. PAB11244
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against synthetic peptide of PIN1.

The human PIN1 gene encodes an essential nuclear peptidylprolyl cis-trans isomerase (PPIase; EC 5.2.1.8) involved in regulation of mitosis. PIN1 belongs to a class of PPIases that includes the E. coli parvulin, yeast Ess1, and Drosophila dodo (dod) gene products (Lu et al., 1996 [PubMed 8606777]).[supplied by OMIM

Specifications

Antigen PIN1
Applications ELISA, Immunoprecipitation, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against synthetic peptide of PIN1.
Dilution ELISA (1:2500-1:10000) Western Blot (1:500-1:3000) The optimal working dilution should be determined by the end user.
Formulation In 20mM KH2PO4, 150mM NaCl, pH 7.2 (0.01% sodium azide)
Gene PIN1
Gene Alias DOD/UBL5
Gene Symbols PIN1
Host Species Rabbit
Immunogen A synthetic peptide corresponding to internal region of human PIN1.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5300
Target Species Bovine, Canine, Human, Monkey, Mouse, Rat
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.