Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pla2g4a, Rabbit, Polyclonal Antibody, Abnova™

Catalog No. 89116784 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-116-784 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-116-784 Supplier Abnova Corporation Supplier No. PAB13651
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against synthetic peptide of Pla2g4a.

Sequence: DQEKTFRQQRKENIKENMKKLLGPKKSE

Specifications

Antigen Pla2g4a
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against synthetic peptide of Pla2g4a.
Dilution Western Blot (1-5 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In PBS (0.05% sodium azide)
Gene Pla2g4a
Gene Alias Pla2g4/cPLA2/cPLA2alpha
Gene Symbols Pla2g4a
Host Species Rabbit
Immunogen A synthetic peptide corresponding to amino acids 152-179 of mouse Pla2g4a.
Quantity 1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 18783
Test Specificity This polyclonal antibody is specific to cytosolic Phospholipase A2 (cPLA2). This antibody against a unique peptide in murine cPLA2. It is very useful for studies of the expression of this arachidonate-specific PLA2 and its role in the regulation of eicosanoid metabolism.
Target Species Mouse
Content And Storage Store at 4°C. For long term storage store at -80°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.