Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Plat Rabbit anti-Mouse, Polyclonal Antibody, Abnova™

Catalog No. 89116787 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-116-787 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-116-787 Supplier Abnova Corporation Supplier No. PAB13664
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against recombinant Plat.

Specifications

Antigen Plat
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against recombinant Plat.
Dilution ELISA (1:100000) Western Blot (1:10000) The optimal working dilution should be determined by the end user.
Formulation In 50mM sodium phosphate buffer, 100mM NaCl, 1mM EDTA, pH 7.4
Gene Plat
Gene Alias AU020998/AW212668/D8Ertd2e/MGC18508/t-PA/tPA
Gene Symbols Plat
Host Species Rabbit
Immunogen Recombinant protein corresponding to mouse Plat.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 18791
Test Specificity These polyclonal antibodies are specific to Mouse tPA. They are available as serum, IgG fraction, or affinity purified as well as labeled with biotin and FITC.
Target Species Mouse
Content And Storage Store at -80°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.