Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PLAU Rabbit anti-Human, Polyclonal Antibody, Abnova™

Catalog No. 89117348 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-117-348 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-117-348 Supplier Abnova Corporation Supplier No. PAB14920
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against native PLAU.

This gene encodes a serine protease involved in degradation of the extracellular matrix and possibly tumor cell migration and proliferation. A specific polymorphism in this gene may be associated with late-onset Alzheimer's disease and also with decreased affinity for fibrin-binding. This protein converts plasminogen to plasmin by specific cleavage of an Arg-Val bond in plasminogen. Plasmin in turn cleaves this protein at a Lys-Ile bond to form a two-chain derivative in which a single disulfide bond connects the amino-terminal A-chain to the catalytically active, carboxy-terminal B-chain. This two-chain derivative is also called HMW-uPA (high molecular weight uPA). HMW-uPA can be further processed into LMW-uPA (low molecular weight uPA) by cleavage of chain A into a short chain A (A1) and an amino-terminal fragment. LMW-uPA is proteolytically active but does not bind to the uPA receptor. Alternatively spliced transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq]

Specifications

Antigen PLAU
Applications ELISA, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Frozen), Immunohistochemistry (PFA fixed), Immunoprecipitation, Inhibition Assays, Radioimmune Assays (RIA), Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against native PLAU.
Dilution The optimal working dilution should be determined by the end user.
Formulation In serum
Gene PLAU
Gene Alias ATF/UPA/URK/UROKINASE/u-PA
Gene Symbols PLAU
Host Species Rabbit
Immunogen Native purified two chain human urinary type high molecular weight PLAU from pooled urine.
Quantity 1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5328
Test Specificity This antibody reacts only with human t-PA. No reactivity with human high molecular weight urokinase. Identity established by double diffusion with t-PA isolated from human, uterine tissue human and CHO derived recombinant t-PA. No other proteins so far tested showed greater than 1% cross reaction.
Target Species Human
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.