Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PLCB3 Rabbit anti-Human, Mouse, Rat, Polyclonal Antibody, Abnova™

Catalog No. 89118946 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-118-946 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-118-946 Supplier Abnova Corporation Supplier No. PAB17281
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against synthetic peptide of PLCB3.

PLCB3 plays an important role in initiating receptor-mediated signal transduction. Activation of PLC takes place in many cells as a response to stimulation by hormones, growth factors, neurotransmitters, and other ligands (Lagercrantz et al., 1995 [PubMed 7607669]).[supplied by OMIM

Specifications

Antigen PLCB3
Applications Immunohistochemistry (PFA fixed), Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against synthetic peptide of PLCB3.
Dilution Westen Blot (1:500-1:1000) Immunohistochemistry (1:50-1:100) ELISA (1:40000) The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.4 (150mM NaCl, 0.02% sodium azide, 50% glycerol)
Gene PLCB3
Gene Alias FLJ37084
Gene Symbols PLCB3
Host Species Rabbit
Immunogen A synthetic peptide corresponding to residues surrounding serine 1105 of human PLCB3.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5331
Test Specificity This antibody detects endogenous levels of total PLCB3 protein.
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.