Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PLG Rabbit anti-Mouse, Polyclonal Antibody, Abnova™

Catalog No. 89116785 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-116-785 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-116-785 Supplier Abnova Corporation Supplier No. PAB13652
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against Plg.

Specifications

Antigen PLG
Applications ELISA, Immunohistochemistry, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against Plg.
Dilution The optimal working dilution should be determined by the end user.
Formulation In 50mM sodium phosphate buffer, 100mM NaCl, 1mM EDTA, pH 6.6
Gene Plg
Gene Alias AI649309/Pg
Gene Symbols Plg
Host Species Rabbit
Immunogen Purified mouse Plg.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 18815
Test Specificity This polyclonal antibody are specific to plasminogen in mouse and are a useful tool in fibrnolytic and cancer studies. This is a high titer preparation. This is available as antiserum, IgG fraction, affinity purified as well as FITC and Biotin labeled.
Target Species Mouse
Content And Storage Store at -80°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.