Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pmaip1 Rabbit anti-Mouse, Polyclonal Antibody, Abnova™

Catalog No. 89114370 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-114-370 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-114-370 Supplier Abnova Corporation Supplier No. PAB0722
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against synthetic peptide of Pmaip1.

Sequence: MPGRKARRNAPVNPTR

Specifications

Antigen Pmaip1
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against synthetic peptide of Pmaip1.
Dilution Western Blot (1:1000-1:2000) The optimal dilution should be determined by the end user.
Formulation Lyophilized from PBS
Gene Pmaip1
Gene Alias Noxa
Gene Symbols Pmaip1
Host Species Rabbit
Immunogen A synthetic peptide (conjugated diphtheria toxoid) corresponding to amino acids 1-16 of mouse Pmaip1.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 58801
Test Specificity Western blot analysis of cells infected with Noxa adenoviruses and BAF indicates a high level of specificity for this antiserum.
Target Species Mouse
Content And Storage Store at 4°C on dry atmosphere. After reconstitution with deionized water store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Lyophilized
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.