Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Rabbit anti-PMEL17/SILV, Clone: P14-V, Novus Biologicals™

Rabbit Monoclonal Antibody
$524.00 - $768.00
Specifications
Antigen | PMEL17/SILV |
---|---|
Clone | P14-V |
Concentration | 0.5mg/mL |
Dilution | Immunohistochemistry 1:100-1:200, Immunohistochemistry-Paraffin 1:100-1:200 |
Applications | Immunohistochemistry, Immunohistochemistry (Paraffin) |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP17900110
![]() |
Novus Biologicals
NBP179001100UL |
100 μL |
Each for $524.00
|
|
|||||
NBP17900150
![]() |
Novus Biologicals
NBP179001500UL |
500 μL |
Each for $768.00
|
|
|||||
Description
PMEL17/SILV Monoclonal specifically detects PMEL17/SILV in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
PMEL17/SILV | |
0.5mg/mL | |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
RUO | |
20mM Tris/HCl (pH 8.0) and 20 mg/ml BSA with 0.05% Sodium Azide | |
6490 | |
Peptide derived from C-terminal sequence of human melanosome/gp10. | |
Affinity Purified |
P14-V | |
Immunohistochemistry 1:100-1:200, Immunohistochemistry-Paraffin 1:100-1:200 | |
Monoclonal | |
Rabbit | |
Human | |
D12S53EP1, gp100, ME20, ME20-M, melanocyte protein mel 17, Melanocyte protein Pmel 17, Melanocytes lineage-specific antigen GP100, Melanoma-associated ME20 antigen, melanosomal matrix protein17, PMEL17P100, premelanosome proteinME20M, SI, SIL, silver (mouse homolog) like, silver homolog (mouse), Silver locus protein homolog, silver, mouse, homolog of, SILVPmel17 | |
PMEL | |
Primary | |
Store at 4C. Do not freeze. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title