Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PML Rabbit anti-Human, Polyclonal Antibody, Abnova™

Catalog No. 89117237 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-117-237 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-117-237 Supplier Abnova Corporation Supplier No. PAB14682
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against partial recombinant PML.

The protein encoded by this gene is a member of the tripartite motif (TRIM) family. The TRIM motif includes three zinc-binding domains, a RING, a B-box type 1 and a B-box type 2, and a coiled-coil region. This phosphoprotein localizes to nuclear bodies where it functions as a transcription factor and tumor suppressor. Its expression is cell-cycle related and it regulates the p53 response to oncogenic signals. The gene is often involved in the translocation with the retinoic acid receptor alpha gene associated with acute promyelocytic leukemia (APL). Extensive alternative splicing of this gene results in several variations of the protein's central and C-terminal regions; all variants encode the same N-terminus. Alternatively spliced transcript variants encoding different isoforms have been identified. [provided by RefSeq]

Specifications

Antigen PML
Applications Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against partial recombinant PML.
Dilution Western Blot (1:5000) Immunofluorescence (1:500) Immunohistochemistry (1:100) The optimal working dilution should be determined by the end user.
Formulation Lyophilized from 10mM PBS, pH 7.4
Gene PML
Gene Alias MYL/PP8675/RNF71/TRIM19
Gene Symbols PML
Host Species Rabbit
Immunogen Recombinant protein corresponding to the ring finger in the homodimerization region of human PML.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5371
Test Specificity This antibody recognizes normal PML proteins of lower molecular weight, and also recognizes the high molecular weight PML/RARalpha chimeric protein in promyelocytic leukemia patients.
Target Species Human
Content And Storage Store at -20°C on dry atmosphere. After reconstitution with appropriate volume of deionized water to reach a final antibody concentration of 1mg/mL store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Lyophilized
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.