Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PODXL Rabbit anti-Human, Biotin, Polyclonal Antibody, Abnova™

Catalog No. 89117556 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-117-556 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-117-556 Supplier Abnova Corporation Supplier No. PAB15301
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against synthetic peptide of PODXL.

This gene encodes a member of the sialomucin protein family. The encoded protein was originally identified as an important component of glomerular podocytes. Podocytes are highly differentiated epithelial cells with interdigitating foot processes covering the outer aspect of the glomerular basement membrane. Other biological activities of the encoded protein include: binding in a membrane protein complex with Na+/H+ exchanger regulatory factor to intracellular cytoskeletal elements, playing a role in hematopoetic cell differentiation, and being expressed in vascular endothelium cells and binding to L-selectin. [provided by RefSeq]

Specifications

Antigen PODXL
Applications Western Blot
Classification Polyclonal
Conjugate Biotin
Description Rabbit polyclonal antibody raised against synthetic peptide of PODXL.
Dilution Western Blot (2 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In PBS
Gene PODXL
Gene Alias Gp200/MGC138240/PC/PCLP
Gene Symbols PODXL
Host Species Rabbit
Immunogen A synthetic peptide corresponding to amino acids 100-200 of human PODXL.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5420
Test Specificity This antibody can be used for Western blot analysis.
Target Species Human
Content And Storage Store at 4°C. Do not freeze.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.